Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NRG1-beta 1/HRG1-beta 1 EGF Domain Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF396NA
This item is not returnable.
View return policy
Description
Neuregulin-1 beta 1/NRG1 beta 1 Polyclonal specifically detects Neuregulin-1 beta 1/NRG1 beta 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.Specifications
Neuregulin-1 beta 1/NRG1 beta 1 | |
Polyclonal | |
Unconjugated | |
NP_039250 | |
NRG1 | |
E. coli-derived recombinant human NRG1-beta 1/HRG1-beta 1 EGF Domain Thr176-Lys246 Accession # NP_039250 | |
100 μg | |
Primary | |
Detects human NRG1-beta 1/HRG1-beta 1 EGF Domain in direct ELISAs and Western blots. In Western blots (reducing conditions), approximately 100% cross-reactivity with recombinant human HRG-alpha is observed. In direct ELISAs, approximately 5% cross-reactivity with recombinant human HRG-alpha is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.5-2.0 ug/mL | |
ARIA, GGF, GGF2, Heregulin-1, HGL, HRG, HRG1, HRG1-alpha, HRG1-beta 1, HRGA, MST131, MSTP131, NDF, neuregulin 1, Neuregulin1, Neuregulin1 beta 1, NRG1, NRG1-IT2, SMDF | |
Goat | |
Affinity Purified | |
RUO | |
3084 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction