Learn More
Human NRG1-beta 1/HRG1-beta 1 Extracellular Domain Biotinylated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | Neuregulin-1 beta 1/NRG1 beta 1 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Biotin |
| Dilution | Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL |
| Gene Accession No. | AAA58639 |
| Gene Alias | ARIA, GGF, GGF2, Heregulin-1, HGL, HRG, HRG1, HRG1-alpha, HRG1-beta 1, HRGA, MST131, MSTP131, NDF, neuregulin 1, Neuregulin1, Neuregulin1 beta 1, NRG1, NRG1-IT2, SMDF |
| Gene Symbols | NRG1 |
| Host Species | Goat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.