Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NRG1 Isoform GGF2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2015
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2015 100 μg
AF2015SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2015 Supplier R&D Systems Supplier No. AF2015
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Neuregulin-1 Isoform GGF2 Polyclonal specifically detects Neuregulin-1 Isoform GGF2 in Human samples. It is validated for Western Blot.

Specifications

Antigen Neuregulin-1 Isoform GGF2
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. NP_039256
Gene Alias GGF2, ARIA, GGF, GGF2, Heregulin-1, HGL, HRG, HRG1, HRG1-alpha, HRG1-beta 1, HRGA, MST131, MSTP131, NDF, Neuregulin1, Neuregulin1 Isoform GGF2, NRG1, NRG1-IT2, SMDF
Gene Symbols NRG1
Host Species Goat
Immunogen E. coli-derived recombinant human NRG1 Isoform GGF2 Gly51-Cys248 Accession # NP_039256
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3084
Test Specificity Detects human NRG1 Isoform GGF2 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) NRG-1 alpha EGF domain, rhNRG1-beta 1 extracellular domain, rhNRG1-beta 1 EGF domain, rhSMDF, and recombinant mouse NRG3 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.