Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NRL Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF2945
This item is not returnable.
View return policy
Description
NRL Polyclonal specifically detects NRL in Human samples. It is validated for Western Blot.Specifications
NRL | |
Polyclonal | |
Unconjugated | |
P54845 | |
NRL | |
E. coli-derived recombinant human NRL Met1-Leu237 Accession # P54845 | |
100 μg | |
Primary | |
Detects human NRL in direct ELISAs and Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
D14S46Eneural retina-specific leucine zipper protein, neural retina leucine zipper, NRL-MAF, RP27, RP27neural retinal-specific leucine zipper | |
Goat | |
Affinity Purified | |
RUO | |
4901 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction