Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NT-3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF267NA
This item is not returnable.
View return policy
Description
NT-3 Polyclonal specifically detects NT-3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.Specifications
NT-3 | |
Polyclonal | |
Unconjugated | |
P20783 | |
NTF3 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human NT-3 Tyr139-Thr257 Accession # P20783 | |
100 μg | |
Primary | |
Detects human NT-3 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human (rh) NT-4 and rhBDNF is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.1-0.4 ug/mL | |
HDNF, MGC129711, Nerve growth factor 2, Neurotrophic factor, neurotrophin 3, neurotrophin-3, NGF2, NGF-2, NT3, NT-3, NTF3 | |
Goat | |
Affinity Purified | |
RUO | |
4908 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction