Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NT-4 Propeptide Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3450
This item is not returnable.
View return policy
Description
NT-4 Polyclonal specifically detects NT-4 in Human samples. It is validated for Western Blot.Specifications
NT-4 | |
Polyclonal | |
Unconjugated | |
P34130 | |
NTF4 | |
E. coli-derived recombinant human NT-4 Propeptide Gln25-Arg80 Accession # P34130 | |
100 μg | |
Primary | |
Detects human NT-4 Propeptide in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with mature recombinant human NT-4 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
GLC10, GLC1O, neurotrophic factor 4, neurotrophic factor 5, neurotrophin 4, neurotrophin 5 (neurotrophin 4/5), neurotrophin-4, neurotrophin-5, Neutrophic factor 4, NT4, NT-4, NT-4/5, NT-5, NTF4, NTF5, NTF5NT5 | |
Goat | |
Affinity Purified | |
RUO | |
4909 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction