Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human NT-ProANP Quantikine ELISA Kit
SDP

Catalog No. DANP00 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DANP00 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DANP00 Supplier R&D Systems™ Supplier No. DANP00

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

E. coli-expressed recombinant human NT-ProANP

For the determination of natural and recombinant NT-ProANP in humans. R&D Systems Human NT-ProANP Quantikine™ ELISA Kit is a 4.5 hour solid-phase ELISA designed to measure NT-ProANP in cell culture supernates, serum and plasma.

The Human NT-ProANP Quantikine ELISA Kit quantifies human Atrial Natriuretic Peptide/ANP in cell culture supernates (50 ul), serum (25 ul), edta plasma (25 ul), heparin plasma (25 ul).

Specifications

Description ANF, ANP, ANPatriopeptin, ATFB6, atrial natriuretic factor, Atrial natriuretic peptide, Cardionatrin, CDD-ANF, natriuretic peptide A, natriuretic peptide precursor A, NPPA, PND, PNDcardionatrin, prepronatriodilatin
Product Type ELISA Kit
Quantity 1 Kit
Species Human
Conjugate HRP
Assay Sensitivity 0.146 ng/mL
Assay Range 0.31 to 20 ng/mL
Cross Reactivity <0.5% cross-reactivity observed with available related molecules, <50% cross-species reactivity observed with species tested
For Use With (Application) ELISA
Gene ID (Entrez) 4878
Product Line R&D Systems
Sample Type Cell Culture Supernates (50uL), Serum (25uL), EDTA Plasma (25uL), Heparin Plasma (25uL)
Sample Volume 25 μL (Serum), 50 μL (Cell Culture Supernates), 25 μL (Heparin plasma), 25 μL (EDTA plasma)
Test Time 4.5 hr.
Synonym ANF, ANP, ANPatriopeptin, ATFB6, atrial natriuretic factor, Atrial natriuretic peptide, Cardionatrin, CDD-ANF, natriuretic peptide A, natriuretic peptide precursor A, NPPA, PND, PNDcardionatrin, prepronatriodilatin
Kit Contents E. coli-expressed recombinant human NT-ProANP
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Research Area Cardiovascular
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.