Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NTAL Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC4066N100UG
This item is not returnable.
View return policy
Description
NTAL Monoclonal specifically detects NTAL in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
NTAL | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9GZY6 | |
LAT2 | |
E. coli-derived recombinant human NTAL Met1-Ala243 Accession # Q9GZY6 | |
RUO | |
7462 | |
Human | |
IgG2b |
Flow Cytometry | |
440005 | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
LAB, LABWilliams-Beuren syndrome chromosomal region 15 protein, Linker for activation of B-cells, linker for activation of T cells family, member 2, linker for activation of T-cells family member 2, Membrane-associated adapter molecule, Non-T-cell activation linker, NTALWilliams-Beuren syndrome chromosomal region 5 protein, WBS15, WBSCR15, WBSCR5, WBSCR5HSPC046, Williams-Beuren syndrome chromosome region 5, WSCR5 | |
Mouse | |
100 μg | |
Primary | |
Detects human NTAL in direct ELISAs and Western blots. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction