Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NTB-A/SLAMF6 Alexa Fluor™ 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FA19081U100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FA19081U100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FA19081U100 Supplier R&D Systems Supplier No. FAB19081U100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

NTB-A/SLAMF6/CD352 Monoclonal specifically detects NTB-A/SLAMF6/CD352 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen NTB-A/SLAMF6/CD352
Applications Flow Cytometry
Classification Monoclonal
Clone 292811
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 350
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q96DU3
Gene Alias Activating NK receptor, CD352, CD352 antigen, KALIFLJ50657, Ly108, NK-T-B-antigen, NTBA, NTBA receptor, NTB-AMGC104953, NTBAT- and B-cell antigen, SF2000, SLAM family member 6, SLAMF6
Gene Symbols SLAMF6
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human NTB-A/SLAMF6 Leu28-Lys225 Accession # Q96DU3
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114836
Test Specificity Detects human NTB-A/SLAMF6 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) BLAME or rhCRACC is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.