Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NTB-A/SLAMF6 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 6 publications
Supplier: R&D Systems MAB19081SP
This item is not returnable.
View return policy
Description
NTB-A/SLAMF6/CD352 Monoclonal specifically detects NTB-A/SLAMF6/CD352 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
NTB-A/SLAMF6/CD352 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Q96DU3 | |
SLAMF6 | |
Mouse myeloma cell line NS0-derived recombinant human NTB-A/SLAMF6 Leu28-Lys225 Accession # Q96DU3 | |
25 μg | |
Primary | |
Detects human NTB-A/SLAMF6 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) BLAME or rhCRACC is observed. | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
292811 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Activating NK receptor, CD352, CD352 antigen, KALIFLJ50657, Ly108, NK-T-B-antigen, NTBA, NTBA receptor, NTB-AMGC104953, NTBAT- and B-cell antigen, SF2000, SLAM family member 6, SLAMF6 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
114836 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction