Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NTB-A/SLAMF6 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1908
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1908 100 μg
AF1908SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF1908 Supplier R&D Systems Supplier No. AF1908
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

NTB-A/SLAMF6/CD352 Polyclonal specifically detects NTB-A/SLAMF6/CD352 in Human samples. It is validated for Western Blot.

Specifications

Antigen NTB-A/SLAMF6/CD352
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q96DU3
Gene Alias Activating NK receptor, CD352, CD352 antigen, KALIFLJ50657, Ly108, NK-T-B-antigen, NTBA, NTBA receptor, NTB-AMGC104953, NTBAT- and B-cell antigen, SF2000, SLAM family member 6, SLAMF6
Gene Symbols SLAMF6
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human NTB-A/SLAMF6 Leu28-Lys225 Accession # Q96DU3
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114836
Test Specificity Detects human NTB-A/SLAMF6 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse NTB-A/SLAMF6 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.