Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human NTB-A/SLAMF6 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB19081A02
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB19081A02 25 Tests
FAB19081A10 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB19081A02 Supplier R&D Systems Supplier No. FAB19081A025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

NTB-A/SLAMF6/CD352 Monoclonal specifically detects NTB-A/SLAMF6/CD352 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen NTB-A/SLAMF6/CD352
Applications Flow Cytometry
Classification Monoclonal
Clone 292811
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q96DU3
Gene Alias Activating NK receptor, CD352, CD352 antigen, KALIFLJ50657, Ly108, NK-T-B-antigen, NTBA, NTBA receptor, NTB-AMGC104953, NTBAT- and B-cell antigen, SF2000, SLAM family member 6, SLAMF6
Gene Symbols SLAMF6
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human NTB-A/SLAMF6 Leu28-Lys225 Accession # Q96DU3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114836
Test Specificity Detects human NTB-A/SLAMF6 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) BLAME or rhCRACC is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.