Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals Human NTB-A/SLAMF6/CD352 ELISA Kit (Colorimetric)
SDP

Catalog No. NB231242 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB231242 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB231242 Supplier Novus Biologicals Supplier No. NBP280354

Specifications

Assay Range 15.63-1000 pg/mL
Assay Sensitivity 12.7 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Recombinant Protein
Specificity Recognizes both recombinant and natural Human NTB-A/SLAMF6/CD352.
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 114836
Gene Symbol SLAMF6
Quantity 1 Kit
Storage Requirements Storage is content dependent.
Synonym Activating NK receptor, CD352 antigen, KALIFLJ50657, Ly108, NK-T-B-antigen, NTBA receptor, NTB-AMGC104953, NTBAT- and B-cell antigen, SF2000, SLAM family member 6
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.