Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NTB-A/SLAMF6 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
$151.20 - $444.00
Specifications
Antigen | NTB-A/SLAMF6/CD352 |
---|---|
Clone | 292811 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB19081P02
![]() |
R&D Systems
FAB19081P025 |
25 Tests |
Each for $151.20
|
|
|||||
FAB19081P10
![]() |
R&D Systems
FAB19081P100 |
100 Tests |
Each for $444.00
|
|
|||||
Description
NTB-A/SLAMF6/CD352 Monoclonal specifically detects NTB-A/SLAMF6/CD352 in Human samples. It is validated for Flow Cytometry.Specifications
NTB-A/SLAMF6/CD352 | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
Activating NK receptor, CD352, CD352 antigen, KALIFLJ50657, Ly108, NK-T-B-antigen, NTBA, NTBA receptor, NTB-AMGC104953, NTBAT- and B-cell antigen, SF2000, SLAM family member 6, SLAMF6 | |
SLAMF6 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human NTB-A/SLAMF6 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) BLAME or rhCRACC is observed. |
292811 | |
Flow Cytometry | |
R-PE | |
Mouse | |
Human | |
Q96DU3 | |
114836 | |
Mouse myeloma cell line NS0-derived recombinant human NTB-A/SLAMF6 Leu28-Lys225 Accession # Q96DU3 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title