Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Nurr1 (aa 4-124) Control Fragment Recombinant Protein

Catalog No. RP93701
Encompass_Preferred
Click to view available options
Quantity:
100 μL

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The Nuclear receptor related 1 protein (Nurr1) also known as NR4A2 (nuclear receptor subfamily 4, group A, member 2) is a protein that in humans is encoded by the NR4A2 gene. NURR1 is a member of the nuclear receptor family of intracellular transcription factors.Nurr1 plays a key role in the maintenance of the dopaminergic system of the brain. Mutations in this gene have been associated with disorders related to dopaminergic dysfunction, including Parkinson's disease, schizophrenia, and manic depression. Misregulation of this gene may be associated with rheumatoid arthritis. Four transcript variants encoding four distinct isoforms have been identified for this gene. Additional alternate splice variants may exist, but their full-length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P43354
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 4929
Name Human Nurr1 (aa 4-124) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias HZF3; Hzf-3; Immediate-early response protein NOT; intermediate-early receptor protein; NGFI-B; NGFI-B beta; NGFI-B/nur77 beta-type transcription factor homolog; NOT; NR4A2; Nuclear orphan receptor HZF-3; nuclear receptor related 1; nuclear receptor subfamily 4 group A member 2; nuclear receptor subfamily 4 group A member 2 variant TINUR; nuclear receptor subfamily 4 group A member 2 variant NURR1a; nuclear receptor subfamily 4 group A member 2 variant NURR1b; nuclear receptor subfamily 4 group A member 2 variant NURR1c; nuclear receptor subfamily 4 group A member 2 variant NURR2c; nuclear receptor subfamily 4, group A, member 2; nur related protein 1; nur related protein-1, human homolog of; Nurr1; NUR-related factor 1; orphan nuclear receptor NR4A2; orphan nuclear receptor NURR1; Regenerating liver nuclear receptor 1; RNR1; RNR-1; SL-322; T-cell nuclear receptor NOT; TINOR; TINUR; transcriptionally inducible nuclear receptor related 1; transcriptionally-inducible nuclear receptor
Common Name Nurr1
Gene Symbol NR4A2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VQAQYGSSPQGASPASQSYSYHSSGEYSSDFLTPEFVKFSMDLTNTEITATTSLPSFSTFMDNYSTGYDVKPPCLYQMPLSGQQSSIKVEDIQMHNYQQHSHLPPQSEEMMPHSGSVYYKP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.