Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human OAS2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1925
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1925 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB1925 Supplier R&D Systems Supplier No. MAB1925
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

OAS2 Monoclonal specifically detects OAS2 in Human samples. It is validated for Immunoprecipitation, Immunocytochemistry.

Specifications

Antigen OAS2
Applications Immunoprecipitation, Immunocytochemistry
Classification Monoclonal
Clone HLS56/3
Conjugate Unconjugated
Dilution Immunoprecipitation, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias (2-5')oligo(A) synthase 2, (2'-5')oligo(A) synthetase 2, (2-5')oligo(A) synthetase 2, 2'5' oligoadenylate synthetase 2, 2-5A synthase 2, 2-5A synthetase 2, 2'-5'-oligoadenylate synthase 2, 2'-5'-oligoadenylate synthetase 2 (69-71 kD), 2'-5'-oligoadenylate synthetase 2, 69/71kDa, EC 2.7.7, EC 2.7.7.-, MGC78578, p69 OAS p71 OAS, p69OAS p71OAS
Gene Symbols OAS2
Host Species Mouse
Immunogen 2Æ,5Æ-Oligoadenylate Synthetase-2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4939
Test Specificity Detects human OAS2.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.