Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human OBCAM/OPCML Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
$200.50 - $549.50
Specifications
Antigen | OBCAM/OPCML |
---|---|
Clone | 341723 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB27771SP
![]() |
R&D Systems
MAB27771SP |
25 μg |
Each for $200.50
|
|
|||||
MAB27771
![]() |
R&D Systems
MAB27771 |
100 μg |
Each for $549.50
|
|
|||||
Description
OBCAM/OPCML Monoclonal specifically detects OBCAM/OPCML in Human samples. It is validated for Western Blot.Specifications
OBCAM/OPCML | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q14982 | |
4978 | |
Mouse myeloma cell line NS0-derived recombinant human OBCAM Gly28-Asn322 Accession # Q14982 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human OBCAM on direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) ALCAM, rhBCAM, rhEpCAM, rhNCAM, recombinant mouse (rm) MAdCAM, rmOCAM, or rmNCAM-L1. |
341723 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
IgLON family member 1, IGLON1opioid-binding protein/cell adhesion molecule-like, OBCAMopiate binding-cell adhesion molecule, OPCM, OPCML, opioid binding protein/cell adhesion molecule-like, opioid binding protein/cell adhesion molecule-like preprotein, Opioid-binding cell adhesion molecule, opioid-binding protein/cell adhesion molecule | |
OPCML | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title