Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human OBSCN Partial ORF (NP_443075, 6521 a.a. - 6620 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89954940
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

The obscurin gene spans more than 150 kb, contains over 80 exons and encodes a protein of approximately 720 kDa. The encoded protein contains 68 Ig domains, 2 fibronectin domains, 1 calcium/calmodulin-binding domain, 1 RhoGEF domain with an associated PH domain, and 2 serine-threonine kinase domains. This protein belongs to the family of giant sacromeric signaling proteins that includes titin and nebulin, and may have a role in the organization of myofibrils during assembly and may mediate interactions between the sarcoplasmic reticulum and myofibrils. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]

Sequence: LGPRGPLGLFRPEPRGASPPGPQVRSLEGTSFLLREAPARPVGSAPWTQSFCTRIRRSADSGQSSFTTELSTQTVNFGTVGETVTLHICPDRDGDEAAQP

Specifications

Accession Number NP_443075
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 84033
Molecular Weight (g/mol) 36.63kDa
Name OBSCN (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen LGPRGPLGLFRPEPRGASPPGPQVRSLEGTSFLLREAPARPVGSAPWTQSFCTRIRRSADSGQSSFTTELSTQTVNFGTVGETVTLHICPDRDGDEAAQP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp666E245/FLJ14124/KIAA1556/KIAA1639/MGC120409/MGC120410/MGC120411/MGC120412/MGC138590/UNC89
Common Name OBSCN
Gene Symbol OBSCN
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.