Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Oct-4A Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC17591R100UG
This item is not returnable.
View return policy
Description
Oct-4A Monoclonal specifically detects Oct-4A in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Oct-4A | |
Monoclonal | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Mouse | |
100 μg | |
Primary | |
Detects human Oct-4A in Western blots. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Flow Cytometry | |
653108 | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
NP_002692 | |
E.coli-derived recombinant human Oct-4A Met1-Glu135 Accession # NP_002692 | |
RUO | |
5460 | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction