Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human OCTN1/SLC22A4 Alexa Fluor™ 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB10985T10
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB10985T10 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB10985T10 Supplier R&D Systems Supplier No. FAB10985T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

OCTN1/SLC22A4 Monoclonal antibody specifically detects OCTN1/SLC22A4 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen OCTN1/SLC22A4
Applications Flow Cytometry
Classification Monoclonal
Clone 892147
Conjugate Alexa Fluor 594
Dilution Flow Cytometry
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide
Gene Accession No. Q9H015
Gene Alias Ergothioneine transporter, ET transporter, ETT, MGC34546, MGC40524, OCTN1integral membrane transport protein, Organic cation/carnitine transporter 1, SLC22A4, solute carrier family 22 (organic cation/ergothioneine transporter), member 4, solute carrier family 22 member 4, UT2H
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human SLC22A4, Accession # Q9H015
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6583
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.