Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Olfactomedin-2/Noelin-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5614SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5614SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5614SP Supplier R&D Systems Supplier No. AF5614SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Olfactomedin-2/Noelin-2 Polyclonal specifically detects Olfactomedin-2/Noelin-2 in Human samples. It is validated for Western Blot.

Specifications

Antigen Olfactomedin-2/Noelin-2
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O95897
Gene Alias neuronal olfactomedin related ER localized protein 2, NOE2, NOE2noelin 2, noelin-2, NOELIN2, NOELIN2_V1, olfactomedin 2, Olfactomedin2, Olfactomedin-2, OlfC, OLFM2, OM2
Gene Symbols OLFM2
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Olfactomedin-2/Noelin-2 Gln21-Pro454 Accession # O95897
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93145
Test Specificity Detects human Olfactomedin-2/Noelin-2 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse (rm) Olfactomedin-1 is observed and less than 5% cross-reactivity with recombinant human Olfactomedin-4 and rmOlfactomedin-L3 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.