Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Olfactomedin-4/OLFM4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB9218100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9218100 100 μg
MAB9218SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9218100 Supplier R&D Systems Supplier No. MAB9218100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

OLFM4 Monoclonal specifically detects OLFM4 in Human samples. It is validated for Western Blot.

Specifications

Antigen OLFM4
Applications Western Blot
Classification Monoclonal
Clone 806305
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q6UX06
Gene Alias Antiapoptotic protein GW112, bA209J19.1, GC1, G-CSF-stimulated clone 1 protein, GW112, GW112hOLfD, hGC-1, KIAA4294, olfactomedin 4, olfactomedin-4, OlfD, OLFM4, OLM4, UNQ362
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Olfactomedin-4/OLFM4 Asp21-Gln510 Accession # Q6UX06
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10562
Test Specificity Detects human Olfactomedin-4/OLFM4 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.