Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Olig 1, 2, 3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2230
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2230 100 μg
MAB2230SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2230 Supplier R&D Systems Supplier No. MAB2230
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Olig1/2/3 Monoclonal specifically detects Olig1/2/3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.

Specifications

Antigen Olig1/2/3
Applications Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 257224
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias BHLHB1, bHLHb6, BHLHB7, bHLHe21, PRKCBP2, RACK17
Host Species Mouse
Immunogen E. coli-derived recombinant human Olig1 aa 1-272
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116448
Test Specificity Detects human Olig 1, 2, 3. In direct ELISAs, this antibody shows approximately 20% cross-reactivity with recombinant human (rh) Olig2 and 50 - 100% cross-reactivity with rhOlig3.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.