Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human OLT-2/TARM1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$188.28 - $472.87
Specifications
| Antigen | OLT-2 |
|---|---|
| Clone | 2564C |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10059SP
![]() |
R&D Systems
MAB10059SP |
25 μg |
Each for $188.28
|
|
|||||
MAB10059100
![]() |
R&D Systems
MAB10059100 |
100 μg |
Each for $472.87
|
|
|||||
Description
OLT-2 Monoclonal specifically detects OLT-2 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| OLT-2 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| OLT2, OSCAR-Like Transcript-2 Protein, T Cell-Interacting, Activating Receptor On Myeloid Cells, T Cell-Interacting, Activating Receptor On Myeloid Cells 1, TARM1, T-Cell-Interacting, Activating Receptor On Myeloid Cells Protein 1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human OLT-2/TARM-1, Met1-Val235, Accession # B6A8C7 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 2564C | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Rabbit | |
| Human | |
| B6A8C7 | |
| 441864 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title