Learn More
Human OMgp Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody
Supplier:  R&D Systems AF1673SP
 View more versions of this product
Catalog No. AF1673SP
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
OMgp Polyclonal specifically detects OMgp in Human samples. It is validated for Western Blot.
Specifications
Specifications
OMgp
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
oligodendrocyte myelin glycoprotein, OMG, OMGPoligodendrocyte-myelin glycoprotein
Goat
Affinity Purified
RUO
4974
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot
LYOPH
Western Blot 0.1 ug/mL
P23515
OMG
Mouse myeloma cell line NS0-derived recombinant human OMgp Ile25-Asn420 Accession # P23515
25 μg
Primary
Detects human OMgp in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 70% cross-reactivity with recombinant mouse OMgp is observed.
Human
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only