Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Omgp ELISA Kit

Catalog No. EH353RB
Encompass_Preferred
Click to view available options
Quantity:
5 x 96 Tests
96 Tests

ELISA

Human Omgp quantitates human Omgp in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Omgp.

Oligodendrocyte myelin glycoprotein (OMG or OMGP) is a cell adhesion molecule contributing to the interactive process required for myelination in the central nervous system. It is also involved in axonogenesis, neuron projection regeneration, and regulation of collateral sprouting of intact axons in response to injury.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number oligodendrocyte myelin glycoprotein; oligodendrocyte-myelin glycoprotein; Omg; Omgp
Assay Range 0.391 to 25 ng/mL
Assay Sensitivity 0.39 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 4974
Gene Symbol OMG
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target OMG
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.