Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Oncostatin M/OSM Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF295
This item is not returnable.
View return policy
Description
Oncostatin M/OSM Polyclonal specifically detects Oncostatin M/OSM in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, ELISA Detection (Matched Antibody Pair).Specifications
Oncostatin M/OSM | |
Polyclonal | |
Biotin | |
P13725 | |
OSM | |
E. coli-derived recombinant human Oncostatin M/OSM Ala26-Arg221 Accession # P13725 | |
50 μg | |
Primary | |
Detects Oncostatin M/OSM in ELISAs and Western blots. In sandwich immunoassays, less than 0.1% cross-reactivity with recombinant mouse OSM, recombinant human (rh) IL-6, rhIL-2, rhG-CSF and rhCNTF is observed. | |
Human | |
IgG |
Western Blot, Flow Cytometry, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
MGC20461, oncostatin M, oncostatin-M | |
Goat | |
Affinity Purified | |
RUO | |
5008 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction