Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ONECUT2/OC-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6294SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6294SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6294SP Supplier R&D Systems Supplier No. MAB6294SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ONECUT2/OC-2 Monoclonal specifically detects ONECUT2/OC-2 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen ONECUT2/OC-2
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 606014
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O95948
Gene Alias one cut domain family member 2, HNF-6B, OC2, OC-2, one cut homeobox 2
Gene Symbols ONECUT2
Host Species Mouse
Immunogen E. coli-derived recombinant human ONECUT2/OC-2 Gln185-Thr326 Accession # O95948
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9480
Test Specificity Detects human ONECUT2/OC-2 in direct ELISAs and Western blots. No cross-reactivity with recombinant human ONECUT is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.