Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human OPA1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$214.00 - $538.00
Specifications
Antigen | OPA1 |
---|---|
Clone | 1284B |
Dilution | Western Blot 0.2 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
MAB9506SP
![]() |
R&D Systems
MAB9506SP |
Unconjugated | 25 μg |
Each for $214.00
|
|
||||
MAB9506100
![]() |
R&D Systems
MAB9506100 |
Unconjugated | 100 μg |
Each for $538.00
|
|
||||
Description
OPA1 Monoclonal specifically detects OPA1 in Human samples. It is validated for Western Blot.Specifications
OPA1 | |
Western Blot 0.2 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
BERHS, EC 3.6.5.5, FLJ12460, KIAA0567dynamin-like 120 kDa protein, mitochondrial, LargeG, lilr3, MGM1, mitochondrial dynamin-like GTPase, MTDPS14, NPG, NPGlargeG, NTG, NTGmitochondrial dynamin-like 120 kDa protein, optic atrophy 1 (autosomal dominant), Optic atrophy protein 1 | |
A synthetic peptide made to an internal region within amino acid residues 500-600 of human OPA1 Accession # O60313 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human, mouse, and rat OPA1 in Western blots. |
1284B | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
O60313 | |
4976 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title