Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Opticin Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $491.00
Specifications
Antigen | Opticin |
---|---|
Clone | 361328 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Opticin Monoclonal specifically detects Opticin in Human samples. It is validated for Western Blot.Specifications
Opticin | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Human | |
Q9UBM4 | |
26254 | |
Mouse myeloma cell line NS0-derived recombinant human Opticin Ala20-Thr332 Accession # Q9UBM4 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Opticin in direct ELISAs and Western blots. In direct ELIAs and Western blots, no cross-reactivity with recombinant mouse Opticin is observed. |
361328 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
oculoglycan, Oculogylcan, OPT, OPTC, opticin | |
OPTC | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title