Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human OR5C1 Full-length ORF (NP_001001923.1, 1 a.a. - 320 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89955224
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. [provided by RefSeq]

Sequence: MNSENLTRAAVAPAEFVLLGITNRWDLRVALFLTCLPVYLVSLLGNMGMALLIRMDARLHTPMYFFLANLSLLDACYSSAIGPKMLVDLLLPRATIPYTACALQMFVFAGLADTECCLLAAMAYDRYVAIRNPLLYTTAMSQRLCLALLGASGLGGAVSAFVHTTLTFRLSFCRSRKINSFFCDIPPLLAISCSDTSLNELLLFAICGFIQTATVLAITVSYGFIAGAVIHMRSVEGSRRAASTGGSHLTAVAMMYGTLIFMYLRPSSSYALDTDKMASVFYTLVIPSLNPLIYSLRNKEVKEALRQTWSRFHCPGQGSQ

Specifications

Accession Number NP_001001923.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 392391
Molecular Weight (g/mol) 61.4kDa
Name OR5C1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MNSENLTRAAVAPAEFVLLGITNRWDLRVALFLTCLPVYLVSLLGNMGMALLIRMDARLHTPMYFFLANLSLLDACYSSAIGPKMLVDLLLPRATIPYTACALQMFVFAGLADTECCLLAAMAYDRYVAIRNPLLYTTAMSQRLCLALLGASGLGGAVSAFVHTTLTFRLSFCRSRKINSFFCDIPPLLAISCSDTSLNELLLFAICGFIQTATVLAITVSYGFIAGAVIHMRSVEGSRRAASTGGSHLTAVAMMYGTLIFMYLRPSSSYALDTDKMASVFYTLVIPSLNPLIYSLRNKEVKEALRQTWSRFHCPGQGSQ
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias OR5C2P/OR9-31/OR9-F
Common Name OR5C1
Gene Symbol OR5C1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.