Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Orexin B/Hypocretin-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB734
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB734 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB734 Supplier R&D Systems Supplier No. MAB734
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Orexin B/Hypocretin-2 Monoclonal specifically detects Orexin B/Hypocretin-2 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen Orexin B/Hypocretin-2
Applications Immunohistochemistry
Classification Monoclonal
Clone 145202
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O43612
Gene Alias HCRT, Hcrt 2, Hcrt2, Hypocretin, Hypocretin (orexin) neuropeptide precursor, Hypocretin 2, Hypocretin B, Hypocretin precursor, Hypocretin2, Hypocretin-2, Orexin, Orexin neuropeptide precursor, Orexin precursor, OrexinB, PPORX, PPOX
Gene Symbols HCRT
Host Species Mouse
Immunogen KLH-conjugated human Orexin B/Hypocretin-2 synthetic peptide Accession # O43612
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3060
Test Specificity Detects human Orexin B/Hypocretin-2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Orexin A is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.