Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ORF73 HHV8 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB10389100
This item is not returnable.
View return policy
Description
ORF73/HHV8 Monoclonal specifically detects ORF73/HHV8 in Human samples. It is validated for Immunohistochemistry.Specifications
ORF73/HHV8 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
935114 | |
Human | |
Purified |
Immunohistochemistry | |
LN53 | |
Immunohistochemistry 5-25 ug/mL | |
HHV 8, HHV8GK18_gp81, Human Herpes Virus 8, Kaposi's sarcoma associated herpes virus, KSHV, LANA1 | |
Recombinant protein corresponding to the latent nuclear antigen-1 molecule of ORF73 HHV8 | |
100 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction