Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human OSCAR Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF2004
This item is not returnable.
View return policy
Description
OSCAR Polyclonal specifically detects OSCAR in Human samples. It is validated for Western Blot.Specifications
OSCAR | |
Polyclonal | |
Unconjugated | |
AAM19095 | |
OSCAR | |
Mouse myeloma cell line NS0-derived recombinant human OSCAR isoform 2 Asp19-Asn229 (Asn158Ser) Accession # AAM19095 | |
100 μg | |
Primary | |
Detects human OSCAR in direct ELISAs and Western blots. In these formats, approximately 25% cross-reactivity with recombinant mouse OSCAR is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
hOSCAR, MGC33613, osteoclast associated receptor OSCAR-S1, osteoclast associated receptor OSCAR-S2, osteoclast associated, immunoglobulin-like receptor, osteoclast-associated immunoglobulin-like receptor, Osteoclast-associated receptor, PIgR-3, PIGR3, Poly-Ig receptor 3, Polymeric immunoglobulin receptor 3 | |
Goat | |
Affinity Purified | |
RUO | |
126014 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction