Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human OSMR beta Antibody, R&D Systems™

Mouse Monoclonal Antibody
$549.50
Specifications
| Antigen | OSMR beta |
|---|---|
| Clone | 469221 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, ELISA, CyTOF-ready, Knockout Validated 0.25 ug/10^6 cells |
| Applications | Western Blot, Flow Cytometry, ELISA, CyTOF, Immunoassay |
Description
OSMR beta Monoclonal specifically detects OSMR beta in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready, Knockout Validated.Specifications
| OSMR beta | |
| LYOPH | |
| Western Blot, Flow Cytometry, ELISA, CyTOF, Immunoassay | |
| Unconjugated | |
| Mouse | |
| Human | |
| IL-31 receptor subunit beta, IL-31R subunit beta, IL-31RB, IL-31R-beta, Interleukin-31 receptor subunit beta, MGC150627, MGC75127, oncostatin M receptor, oncostatin-M specific receptor beta subunit, oncostatin-M-specific receptor subunit beta, OSM R beta, OSMR, OSMRB, OSMRBMGC150626, PLCA1 | |
| OSMR | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human OSM R beta in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse OSM R beta is observed. |
| 469221 | |
| Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, ELISA, CyTOF-ready, Knockout Validated 0.25 ug/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q99650 | |
| 9180 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title