Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human OSMR beta Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF4389
This item is not returnable.
View return policy
Description
OSMR beta Polyclonal specifically detects OSMR beta in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready, Knockout Validated.Specifications
OSMR beta | |
Polyclonal | |
Unconjugated | |
Q99650 | |
OSMR | |
Mouse myeloma cell line NS0-derived recombinant human OSM R beta Glu28-Ser739 Accession # Q99650 | |
100 μg | |
Primary | |
Detects human OSM R beta in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with recombinant mouse OSM R beta is observed. | |
Human | |
IgG |
Western Blot, Flow Cytometry, CyTOF, KnockDown | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready, Knockout Validated 0.25 ug/10^6 cells | |
IL-31 receptor subunit beta, IL-31R subunit beta, IL-31RB, IL-31R-beta, Interleukin-31 receptor subunit beta, MGC150627, MGC75127, oncostatin M receptor, oncostatin-M specific receptor beta subunit, oncostatin-M-specific receptor subunit beta, OSM R beta, OSMR, OSMRB, OSMRBMGC150626, PLCA1 | |
Goat | |
Affinity Purified | |
RUO | |
9180 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction