Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Osteoactivin/GPNMB Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FA25501T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FA25501T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FA25501T100 Supplier R&D Systems Supplier No. FAB25501T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Osteoactivin/GPNMB Monoclonal specifically detects Osteoactivin/GPNMB in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen Osteoactivin/GPNMB
Applications Flow Cytometry
Classification Monoclonal
Clone 303822
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. NP_002501
Gene Alias DC-HIL, glycoprotein (transmembrane) nmb, glycoprotein NMB, GPNMB, HGFIN, HGFINglycoprotein nmb-like protein, NMBosteoactivin, Transmembrane glycoprotein HGFIN, transmembrane glycoprotein NMB
Gene Symbols GPNMB
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Osteoactivin/GPNMB isoform 2 Ala22-Asn486 Accession # NP_002501
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10457
Test Specificity Detects human Osteoactivin/GPNMB in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse Osteoactivin is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.