Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Osteoadherin/OSAD Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB2884
This item is not returnable.
View return policy
Description
Osteoadherin/OSAD/OMD Monoclonal specifically detects Osteoadherin/OSAD/OMD in Human samples. It is validated for Immunocytochemistry.Specifications
Osteoadherin/OSAD/OMD | |
Monoclonal | |
LYOPH | |
Immunocytochemistry 8-25 ug/mL | |
Q99983 | |
OMD | |
HEK293 human embryonic kidney cell line transfected with human Osteoadherin/OSAD Gln21-Gln421 Accession # Q99983 | |
100 μg | |
Primary | |
Detects human Osteoadherin/OSAD in ELISA. | |
Human | |
Purified |
Immunocytochemistry | |
806001 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Keratan sulfate proteoglycan osteomodulin, KSPG osteomodulin, OMD, OSAD, Osteoadherin, osteoadherin proteoglycan, osteomodulin, SLRR2C, SLRR2Costeoadherin | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4958 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction