Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Osteocalcin Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC113272T100UG
This item is not returnable.
View return policy
Description
Osteocalcin Monoclonal antibody specifically detects Osteocalcin in Human samples. It is validated for Flow CytometrySpecifications
Osteocalcin | |
Monoclonal | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
BGLAP, BGP, bone gamma-carboxyglutamate (gla) protein, bone gamma-carboxyglutamate (gla) protein (osteocalcin), Bone Gla protein, Gamma-carboxyglutamic acid-containing protein, OC, OCN, osteocalcin | |
Osteocalcin/KLH containing peptide, Accession # P02818 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
582014 | |
Intracellular Staining by Flow Cytometry | |
P02818 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
632 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction