Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Osteocrin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB9669100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9669100 100 μg
MAB9669SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9669100 Supplier R&D Systems Supplier No. MAB9669100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Osteocrin Monoclonal specifically detects Osteocrin in Human samples. It is validated for ELISA.

Specifications

Antigen Osteocrin
Applications ELISA
Classification Monoclonal
Clone 311334
Conjugate Unconjugated
Dilution ELISA
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Gene Accession No. P61366
Gene Alias MUSCLIN, osteocrin, OSTN
Host Species Rat
Immunogen E. coli-derived recombinant human Osteocrin Val28-Gly133 Accession # P61366
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 344901
Test Specificity Detects human Osteocrin in direct ELISAs.
Reconstitution Reconstitute at 0.5mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.