Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Osteopontin/OPN Antibody, R&D Systems™

Mouse Monoclonal Antibody
$241.00 - $561.00
Specifications
Antigen | Osteopontin/OPN |
---|---|
Clone | 223112R |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB14332RSP
![]() |
R&D Systems
MAB14332RSP |
25 μg |
Each for $241.00
|
|
|||||
MB14332R100
![]() |
R&D Systems
MAB14332R100 |
100 μg |
Each for $561.00
|
|
|||||
Description
Osteopontin/OPN Monoclonal specifically detects Osteopontin/OPN in Human samples. It is validated for ELISA.Specifications
Osteopontin/OPN | |
ELISA | |
Monoclonal | |
Supernatant | |
Human | |
NP_000573.1 | |
6696 | |
IgG2b | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
223112R | |
ELISA | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
BNSP, Bone sialoprotein 1, Eta-1, MGC110940, Nephropontin, OPN, osteopontin, secreted phosphoprotein 1bone sialoprotein I, early T-lymphocyteactivation 1), secreted phosphoprotein-1 (osteopontin, bone sialoprotein), Spp1, SPP-1, SPP1/CALPHA1 fusion, Urinary stone protein, uropontin | |
Mouse myeloma cell line NS0-derived recombinant human Osteopontin/OPN, Ile17-Asn300, Accession # NP_000573.1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Osteopontin/OPN in ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title