Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Osteoprotegerin/TNFRSF11B Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8051SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8051SP 25 μg
MAB8051100 100 μg
MAB8051500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB8051SP Supplier R&D Systems Supplier No. MAB8051SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 5 publications

Osteoprotegerin/TNFRSF11B Monoclonal specifically detects Osteoprotegerin/TNFRSF11B in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).

Specifications

Antigen Osteoprotegerin/TNFRSF11B
Applications ELISA
Classification Monoclonal
Clone 69127
Conjugate Unconjugated
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O00300
Gene Alias OCIF, OCIFMGC29565, OPG, OPGtumor necrosis factor receptor superfamily member 11B, Osteoclastogenesis inhibitory factor, Osteoprotegerin, TNFRSF11B, TR1, tumor necrosis factor receptor superfamily, member 11b
Gene Symbols TNFRSF11B
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Osteoprotegerin/TNFRSF11B Glu22-Leu401 Accession # O00300
Purification Method Protein A or G purified from ascites
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4982
Test Specificity Detects human Osteoprotegerin/TNFRSF11B in ELISAs. It has been reported (1), and R&D Systems testing has confirmed, that this antibody shows strong cross-reactivity with an unrelated protein, human Carbonic Anhydrase II. In Western blots, this antibody does not cross-react with recombinant human (rh) 4-1BB, rhCD27, rhCD30, recombinant mouse (rm) CD30, rmCD40, rhDR3, rhDR6, rhFas, rmFas, rhGITR, rmGITR, rhHVEM, rhLT beta R, rhNGF R, rmOPG, rhTNF RI, rmTNF RI, rhTNF RII, or rmTNF RII.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.