Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Osterix/Sp7 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7547
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7547 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7547 Supplier R&D Systems Supplier No. MAB7547
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Osterix/Sp7 Monoclonal specifically detects Osterix/Sp7 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen Osterix/Sp7
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 764704
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8TDD2
Gene Alias MGC126598, OSX, OSXosterix, Sp7, Sp7 transcription factor, transcription factor Sp7, Zinc finger protein osterix
Gene Symbols SP7
Host Species Mouse
Immunogen E. coli-derived recombinant human Osterix/Sp7 Met19-Leu288 Accession # Q8TDD2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121340
Test Specificity Detects human Osterix/Sp7 in direct ELISAs.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.