Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Osterix/Sp7 MAb (Clone 764746), R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB75471100
This item is not returnable.
View return policy
Description
Osterix/Sp7 Monoclonal antibody specifically detects Osterix/Sp7 in Human samples. It is validated for Western BlotSpecifications
| Osterix/Sp7 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| MGC126598, OSX, OSXosterix, Sp7, Sp7 transcription factor, transcription factor Sp7, Zinc finger protein osterix | |
| E. coli-derived recombinant human Osterix/Sp7, Met19-Leu288, Accession # Q8TDD2 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 764746 | |
| Western Blot 1 ug/mL | |
| Q8TDD2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 121340 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction