Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Otx2 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Catalog No. IC1979G025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC1979G025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC1979G025 Supplier R&D Systems Supplier No. IC1979G025
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

Otx2 Monoclonal specifically detects Otx2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen Otx2
Applications Flow Cytometry
Classification Monoclonal
Clone 246826
Conjugate Alexa Fluor 488
Dilution Intracellular Staining by Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P32243
Gene Alias homeobox protein OTX2, MCOPS5, MGC45000, orthodenticle homeobox 2, Orthodenticle homolog 2, orthodenticle homolog 2 (Drosophila), OTX2
Gene Symbols OTX2
Host Species Mouse
Immunogen E. coli-derived recombinant human Otx2 (isoform b) Met1-Leu289 Accession # P32243
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5015
Test Specificity Detects human Otx2 in flow cytometry.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.