Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human OX40/TNFRSF4 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FB10543R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FB10543R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FB10543R100 Supplier R&D Systems Supplier No. FAB10543R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

OX40/TNFRSF4 Monoclonal specifically detects OX40/TNFRSF4 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen OX40/TNFRSF4
Applications Flow Cytometry
Classification Monoclonal
Clone 977974
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P43489
Gene Alias ACT-135, ACT35 antigen, ACT35ATC35 antigen, CD134, CD134 antigen, Ly-70, OX40 cell surface antigen, OX40 homologue, OX40L receptor, OX40lymphoid activation antigene ACT35, TAX transcriptionally-activated glycoprotein 1 receptor, tax-transcriptionally activated glycoprotein 1 receptor, TNFRSF4, tumor necrosis factor receptor superfamily member 4, tumor necrosis factor receptor superfamily, member 4, Txgp1, TXGP1L, TXGP1LOX40 antigen
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human OX40/TNFRSF4, Leu29-Ala216, Accession No. P43489
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 7293.0
Test Specificity Detects human OX40/TNFRSF4 in direct ELISAs
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.