Learn More
Novus Biologicals™ Human Oxidized LDL Autoantibody ELISA Kit (Colorimetric)
Shop All R&D Systems Products

Specifications
Specifications
| Assay Sensitivity | 0.56 mIU/mL |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Other Biological Fluids, Plasma, Serum |
| Specificity | This kit recognizes Human Oxidized LDL Autoantibodyin samples. No Significant cross-reactivity or interference between Human Oxidized LDL Autoantibodyand analogues was observed. This kit does not detect a specific autoantibody isotype. |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Measuring Range | 0.94 to 60 mIU/mL |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Target | Oxidized LDL Autoantibody |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.