Learn More
Human P-Cadherin Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Supplier:  R&D Systems FAB861RR100UG
Catalog No. FAB861RR100
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
P-Cadherin Monoclonal specifically detects P-Cadherin in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
P-Cadherin
Monoclonal
Alexa Fluor 647
Supplied 0.2 mg/ml in a saline solution containing BSA and Sodium Azide.
CAD3, cadherin 3, P-cadherin (placental), cadherin 3, type 1, P-cadherin (placental), cadherin-3, CDH3, CDHP, CDHPcalcium-dependent adhesion protein, placental, HJMD, PCAD, PCadherin, PCADP-cadherin, Placental cadherin
Mouse myeloma cell line NS0-derived recombinant human P-Cadherin Asp108-Gly654 Accession # CAA45177
100 μg
1001
Human
Supernatant
Flow Cytometry
104805R
Flow Cytometry 0.25-1 μg/10^6 cells
CAA45177
Mouse
Protein A or G purified from cell culture supernatant
Primary
Detects human P-Cadherin in direct ELISAs.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.