Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human P-Cadherin Antibody, R&D Systems™

Mouse Monoclonal Antibody
$214.00 - $549.50
Specifications
Antigen | P-Cadherin |
---|---|
Clone | 104805R |
Dilution | Western Blot 1μg/mL, Flow Cytometry 0.25μg/106 cells, Immunocytochemistry 5-25μg/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8μg/mL |
Applications | CyTOF, ELISA, Flow Cytometry, Immunocytochemistry, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB861R-SP
![]() |
R&D Systems
MAB861RSP |
25 μg |
Each for $214.00
|
|
|||||
MAB861R-100
![]() |
R&D Systems
MAB861R100 |
100 μg |
Each for $549.50
|
|
|||||
Description
P-Cadherin Monoclonal specifically detects P-Cadherin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).Specifications
P-Cadherin | |
Western Blot 1μg/mL, Flow Cytometry 0.25μg/106 cells, Immunocytochemistry 5-25μg/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8μg/mL | |
Monoclonal | |
Supernatant | |
Human | |
CAA45177 | |
1001 | |
IgG1 | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
104805R | |
CyTOF, ELISA, Flow Cytometry, Immunocytochemistry, Western Blot | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
CAD3, cadherin 3, P-cadherin (placental), cadherin 3, type 1, P-cadherin (placental), cadherin-3, CDH3, CDHP, CDHPcalcium-dependent adhesion protein, placental, HJMD, PCAD, PCadherin, PCADP-cadherin, Placental cadherin | |
Mouse myeloma cell line NS0-derived recombinant human P-Cadherin Asp108-Gly654 Accession # CAA45177 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects human P-Cadherin in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title