Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human P-Cadherin Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF861
This item is not returnable.
View return policy
Description
P-Cadherin Polyclonal specifically detects P-Cadherin in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, CyTOF-ready.Specifications
P-Cadherin | |
Polyclonal | |
Unconjugated | |
CAA45177 | |
CDH3 | |
Mouse myeloma cell line NS0-derived recombinant human P-Cadherin Asp108-Gly654 Accession # CAA45177 | |
100 μg | |
Primary | |
Detects human P-Cadherin in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) Cadherin-11, rhCadherin-12, rhCadherin-13, rhE-Cadherin and less than 1% cross-reactivity with rhCadherin-8, rhCadherin-17, rhN-Cadherin, rhK-Cadherin and recombinant mouse P-Cadherin is observed. | |
Human | |
IgG |
Western Blot, Flow Cytometry, CyTOF | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
CAD3, cadherin 3, P-cadherin (placental), cadherin 3, type 1, P-cadherin (placental), cadherin-3, CDH3, CDHP, CDHPcalcium-dependent adhesion protein, placental, HJMD, PCAD, PCadherin, PCADP-cadherin, Placental cadherin | |
Goat | |
Affinity Purified | |
RUO | |
1001 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction